Cole-Parmer Adapter, Metrohm meter to BNC electrode
Representative image only.
Cole-Parmer Item # EW-05983-70
$66.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdapter, Metrohm meter to BNC electrode
More About this Item
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.