Cole-Parmer Adapter, Radiometer meter to BNC electrode
Representative image only.
Cole-Parmer Item # EW-05983-90
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdapter, Radiometer meter to BNC electrode
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.