Cole-Parmer ADCF Female Luer to 1/8" L Barb Adapter, Polypropylene, 25/pk
Representative image only.
Cole-Parmer Item # EW-30800-08
$10.75USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$10.75USD / Pkg of 25
  • 10-49$9.68USD / Pkg of 25
  • 50-99$9.14USD / Pkg of 25
  • 100+$8.60USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Hose barb
  • Barb Size-Port 2 (in)0
  • DescriptionADCF Female Luer to 1/8" L Barb Adapter, PP, 25/pk
Certificate of Compliance
More About this Item
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Cole-Parmer ADCF Female Luer to 1/8" L Barb Adapter, PP, 25/pk
5.0
(based on 2 reviews)
Reviewed by 2 customers
Displaying reviews 1-2
Back to top
5.0
Great
By 
from Rutland, VT
Verified Buyer
Comments about Cole-Parmer ADCF Female Luer to 1/8" L Barb Adapter, PP, 25/pk:
it fit perfect
5.0
I had lost hope to find it, then .......
By 
from Charlemagne, QC, Canada
Verified Reviewer
Pros
  • Delivery was fast
  • They Are Hard To Find
Cons
    Best Uses
      Comments about Cole-Parmer ADCF Female Luer to 1/8" L Barb Adapter, PP, 25/pk:
      My bipap machine needs two Luer connections to monitor the pressure. Both of them broke and I could not find replacements (even at the manufacturer).My sleep apnea is severe and I considered buying a new machine at approximately 7500 $. Then I found that I could buy 25 replacements for those Luer valves for less than 12 $ at Cole-Parmer.Almost a miracle !
      • Was this a gift?:
      • No
      Displaying reviews 1-2
      Back to top
      Q&A
      Do you have questions about this product?
      Get answers from real customers and in-house experts with AnswerBox.
      2 Questions | 2 Answers
      Displaying questions 1-2
      • Q:
        Do you have an alternative part number that is the same, but autoclaveable?
        Asked on 2/4/2016 by JK from Poland
        1 answer
        • STAFF EXPERT
          A:
          Please see items 45502-04 and 45512-04 which are autoclavable. You must also consider compatibility of these materials with your solution.
          Answered on 2/4/2016 by Polo from Technical Support
      • Q:
        Why not not recommended for autoclaving?
        Asked on 2/6/2015 by Ali
        1 answer
        • STAFF EXPERT
          A:
          There are different types of PP resins; this particular resin has properties which are not conducive to autoclaving.
          Answered on 2/6/2015 by Dan from Technical Support
      Displaying questions 1-2
      Do you have a question about this product? 