Cole-Parmer Adjustable-Volume Pipettor, 100 to 1000 µL
Representative image only.
Cole-Parmer Item # EW-24501-08
Ergonomic finger grip for comfortable use
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
  • Volume: 100 to 1000 µL
  • 3 year warranty
PRODUCT OPTIONS
$170.50USD / Each
$94.50 - $170.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Max Volume (µL)1000
  • Min Volume (µL)100
  • Increments (µL)5
  • Accuracy (%)0.6
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeBlue
  • DescriptionAdjustable-Volume Pipettor, 100 to 1000 µL; 1/EA
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO 9001:2008 Certified Supplier
Key Features
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
  • Volume: 100 to 1000 µL
  • 3 year warranty
More About this Item
These pipettors are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.