Cole-Parmer All-in-One pH/ATC Probe, Sealed/DJ/Epoxy/Tuff-Tip; Corning
Representative image only.
Cole-Parmer Item # EW-27507-24
Save money without giving up temperature compensation!
  • Electrodes designed to work with meters you already have
  • Find the design that works best for your application without losing automatic temperature compensation
  • 1 year warranty
PRODUCT OPTIONS
$218.70USD / Each
$243.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)80
  • ATC ConnectorRCA Plug
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionAll-in-One pH/ATC Probe, Sealed/DJ/Epoxy/Tuff-Tip; Corning
  • Warranty1 year
1 Year Warranty
Key Features
  • Electrodes designed to work with meters you already have
  • Find the design that works best for your application without losing automatic temperature compensation
  • 1 year warranty
More About this Item
“All-in-One” electrodes offer versatility and customization. These electrodes combine the pH reference, pH half-cell, and temperature sensor in one 12mm-diameter electrode body. The seven designs allow you to select between low-maintenance, sealed electrodes or clog-resistant refillable electrodes. Tuff-Tip electrodes feature a semi-domed tip which protects the pH bulb and ensures a quick response in harsh and dirty samples. PBR-style electrodes provide predictable, reliable, and quick responses—no matter what the sample temperature. This design virtually eliminates drift, accuracy problems, and premature performance losses due to temperature. All electrodes include 3-ft cable and BNC connector for pH, except where noted. Corning-compatible electrodes use the same temperature compensation element as Beckman, but offer an RCA plug for the temperature connection.