Cole-Parmer All-in-One pH/ATC Probe, Sealed/SJ/Epoxy; for Series
Representative image only.
Cole-Parmer Item # EW-59002-72
  • 1 year warranty
PRODUCT OPTIONS
$133.20USD / Each
$148.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSealed
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Temperature CompensationYes
  • ATC ConnectorAudio plug, 3.5 mm
  • ConnectorBNC
  • DescriptionAll-in-One pH/ATC Probe, Sealed/SJ/Epoxy; for Series
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use to replace pH/ATC electrodes for discontinued Oakton pH 10, 100, 200, 500, 1000, 2000, 2500 series meters.