Cole-Parmer Ammonia (NH3 as N) 1000 ppm, ISE Calibration Solution
Representative image only.
Cole-Parmer Item # EW-35803-01
Solutions needed for accurate measurements
  • Multiple ion solutions are available
  • 1 year warranty
PRODUCT OPTIONS
$68.50USD / Each
Include Electrodes Today!
Oakton® Ion Selective Electrodes
$230.00 - $649.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Reference Electrolyte1,000 ppm & 10,000 ppm NH4+
  • Ion-Selective ElectrodeAmmonia
  • DescriptionAmmonia (NH3 as N) ISE Calibration Solution
  • Warranty1 year
1 Year Warranty
Key Features
  • Multiple ion solutions are available
  • 1 year warranty
More About this Item
Choose the compatible solution kit to operate your Cole-Parmer ion-selective electrodes properly. Kits contain a 60 mL bottle of fill solution, a 475 mL bottle of standard, and a 475 mL bottle of ISA.
Individual bottles also listed.