Cole-Parmer APC1589 Amber VOA Vials, 40 mL, Open-Top Cap, 400 mg Trisodium Phosphate; 72/Cs

Representative image only.
Cole-Parmer Item # EW-35205-84
Minimize hazardous chemical exposure with pre-filled vials
  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"

PRODUCT OPTIONS

$50.50USD / Case of 144

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)40
  • Width (")1
  • Tube MaterialBorosilicate glass
  • Height (")3
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Cap TypeOpen-Top/Septa
  • Cap Diameter (mm)24
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • SterileNo
  • AutoclavableNo
  • Outside Diameter (in)1
  • Outside Diameter (mm)27.69
  • Fill400 mg Trisodium Phosphate
  • Level1
  • Special Features
  • DescriptionAmber VOA Vials, 40 mL, Open-Top Cap, 400 mg Trisodium Phosphate; 72/Cs

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat