Environmental Express APC1612 Clear VOA Vials, 40 mL, Closed-Top, 25 mL Methanol; 20/Cs

Representative image only.
Environmental Express Item # EW-35205-97
Minimize hazardous chemical exposure with pre-filled vials
  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (in)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeClosed-Top
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Thread24 mm
  • SterileNo
  • AutoclavableNo
  • Outside Diameter (in)1 1/8
  • Outside Diameter (mm)27.69
  • Fill25 mL Methanol
  • Level1
  • Special Features
  • DescriptionClear VOA Vials, 40 mL, Closed-Top, 25 mL Methanol; 20/Cs

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat