Cole-Parmer APC1636 Amber VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized Water; 72/Cs

Representative image only.
Cole-Parmer Item # EW-35207-09
Minimize hazardous chemical exposure with pre-filled vials
  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • Tare weight listed on the label for efficiency
  • Stir bar already inserted in the vial for easy mixing without chemical exposure

PRODUCT OPTIONS

$138.25USD / Case of 72

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)40
  • Width (")1
  • Tube MaterialBorosilicate glass
  • Height (")3
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Cap TypeOpen-Top/Septa
  • Cap Diameter (mm)24
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Septum Type0.125" thick standard septa
  • SterileNo
  • AutoclavableNo
  • Height (in)3
  • Outside Diameter (in)1
  • Height (mm)98.81
  • Outside Diameter (mm)27.69
  • Fill5 mL deionized (DI) water
  • Level1
  • Special FeaturesContains stir bar; tare weight listed on label
  • DescriptionAmber VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized Water; 72/Cs

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • Tare weight listed on the label for efficiency
  • Stir bar already inserted in the vial for easy mixing without chemical exposure

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat