Cole-Parmer Autoradiography Cassette, Aluminum, 10 x 12"
Representative image only.
Cole-Parmer Item # EW-21700-37
Get the results you need affordably
  • Constructed from lightweight aluminum and nylon
  • Special curved door gives exceptional screen-film contact
PRODUCT OPTIONS
$283.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum and Nylon
  • Length (InchShort)10
  • Width (InchShort)12
Key Features
  • Constructed from lightweight aluminum and nylon
  • Special curved door gives exceptional screen-film contact
More About this Item
Aluminum Cassettes have a push-button front for easy use. The lightweight cassettes feature a curved door with a nylon-covered foam cushion to provide exceptional screen contact.