Cole-Parmer Battery-Powered Liquid Level Alarm/Monitor
Representative image only.
Cole-Parmer Item # EW-43301-15
Inexpensive monitor for unattended filling and fluid transfer applications
  • Alarm sounds and LED flashes when liquid contacts the sensor.
  • Place sensor near overflow tube or in tank or drum to guard against overfilling; place near pump or on floor to detect leaks and spills.
  • Measures a compact 1-13/16” x 3” x ¾”; weighs just 2-1/2 ounces.
$59.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VDC)12
  • DescriptionBattery-Powered Liquid Level Alarm/Monitor
Key Features
  • Alarm sounds and LED flashes when liquid contacts the sensor.
  • Place sensor near overflow tube or in tank or drum to guard against overfilling; place near pump or on floor to detect leaks and spills.
  • Measures a compact 1-13/16” x 3” x ¾”; weighs just 2-1/2 ounces.
More About this Item
What's included: sensor with 10-ft (3 m) cable, suction cup for mounting sensor, AA alkaline battery.