Cole-Parmer Binocular Stereozoom Microscope, 10x-40x, boom stand
Representative image only.
Cole-Parmer Item # EW-48920-10
Stereozoom optics let you easily focus through different magnification ranges
  • Head rotates 360°> for different viewing configurations
PRODUCT OPTIONS
$1,070.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x to 40x
  • Maximum Magnification40x
  • Head ConfigurationBinocular
  • Objectives1x to 4x
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Stand TypeUniversal
  • DescriptionBinocular Stereozoom Microscope, 10x-40x, boom stand; 115V
Key Features
  • Head rotates 360°> for different viewing configurations
More About this Item
Stereozoom microscopes feature stepless magnification through zoom control on viewing head. Head rotates 360°>; widefield 10x eyepieces feature adjustable diopter and interpupillary distance for comfortable viewing.
Universal stand does not include a light source; order halogen task lamp 41310-10 at right. Includes a 6-ft cord with three-prong plug.