Cole-Parmer Essentials Black Rubber Stoppers, Assorted Standard Sizes 00-8; 107/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-62990-90
$180.00USD / Pkg of 107
BUY MORE & SAVE
  • 1-5$180.00USD / Pkg of 107
  • 6-29$162.00USD / Pkg of 107
  • 30-119$153.00USD / Pkg of 107
  • 120+$144.00USD / Pkg of 107
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number00-8
  • ColorBlack
  • MaterialRubber
  • DescriptionBlack Rubber Stoppers, Assorted Standard Sizes 00-8; 107/Pk
More About this Item
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.