Cole-Parmer Blind Caps; port size: 12 mm (Stainless Steel)
Representative image only.
Applikon Item # EW-01919-48
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Connections12 mm port
  • DescriptionBlind Caps; port size: 12 mm (Stainless Steel)
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Blind Caps cover unused ports. Come complete with O-rings.