Environmental Express BPC1170U Pre-Cleaned Wide-Mouth Round Bottle, HDPE, Level 1, Cap without Liner, 1 L; 12/CS
Representative image only.
Environmental Express Item # EW-35207-52
Won't break like glass
  • Provide easy-access wide-mouth
  • Lightweight design with leakproof cap
  • Model number BPC1170U
PRODUCT OPTIONS
$25.00USD / Case of 72
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • Capacity (L)1
  • Capacity (oz)32
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene without liner
  • Cap / Stopper Size63 mm
  • Cap ColorWhite
  • Bottle ColorNatural
  • Level Number1
  • Special Features63 mm cap without liner
  • AutoclavableNo
  • DescriptionPre-Cleaned Wide-Mouth Round Bottle, HDPE, Level 1, Cap without Liner, 1 L; 12/Cs
Key Features
  • Provide easy-access wide-mouth
  • Lightweight design with leakproof cap
  • Model number BPC1170U
More About this Item
Choose between Level 1 or 3 pre-cleaned round wide-mouth bottles. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory. Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis. They are ideal for general-use.