Cole-Parmer Bronze Close-Coupled Gear Pump System; Relief/1.0 GPM/TEFC
Representative image only.
Cole-Parmer Item # EW-70738-60
Rugged corrosion-resistant bronze construction provides long service life
  • Compact footprint with close-coupled design
  • Easy assembly to NEMA and IEC motor frames
PRODUCT OPTIONS
$577.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Flow Rate (mL/rev)2.2
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPM)1
  • Suction Lift, Wet (in H₂O)240
  • Suction Lift, Wet (mm of H₂O)6096
  • Max Motor RPM (rpm)1750
  • Motor Enclosure TypeODP
  • Power (VAC)115 / 208 to 230
  • Power (Hz)60
  • Wetted PartsStainless Steel
  • Body MaterialBronze
  • Gear MaterialBronze
  • Inlet Connection (NPT(F))1/4
  • Outlet Connection (NPT(F))1/4
  • Relief ValveYes
  • Max Temperature (° F)140
  • Min Temperature (° F)32
  • Max Temperature (° C)60
  • Min Temperature (° C)0
  • Motor hp1/4
  • DescriptionBronze Close-Coupled Gear Pump System; Relief/1.0 GPM/TEFC
Key Features
  • Compact footprint with close-coupled design
  • Easy assembly to NEMA and IEC motor frames
More About this Item
These rotary gear pumps are suitable for all liquids that are compatible with bronze—most common liquids are water, oil, and mild chemicals (pH from 4 to 11). Long service life is achieved through thoughtful design and chosen materials of construction: bronze pump housings and gears, 303
stainless steel shafts, carbon-graphite bearings, and Buna N lip seal.
Choose a gear pump head to mount to your own motor (adapters for NEMA or IEC motors are sold separately below) or select a complete system with the pump head conveniently mounted to a standard (NEMA 56C) TEFC motor.