This item is now discontinued. Please browse similar items

Cole-Parmer Calcium Chloride ACS desiccant, 500 g

Representative image only.
Cole-Parmer Item # EW-88403-21

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ClassReagent Salts
  • ChemistrySolutions
  • FormSolid
  • Weight (g)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number10043-52-4
  • Shelf Life (months)36
  • DescriptionCalcium Chloride ACS desiccant, (500 g)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat