Cole-Parmer Carboy Washing Kit; 22 inch Brushes, Size 9 Gloves, Micro-90 Cleaner
Representative image only.
Cole-Parmer Item # EW-25000-48
Bundled for convenience and to save you money!
  • We've combined our best-selling brushes, gloves, and cleaner for your carboy cleaning needs.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • ApplicationsManual or ultrasonic; removes most biological materials; cleans glass, most metals, plastic and rubber
  • DescriptionCarboy Washing Kit; 22 inch Brushes, Size 9 Gloves, Micro-90 Cleaner
Key Features
  • We've combined our best-selling brushes, gloves, and cleaner for your carboy cleaning needs.
More About this Item
Each kit comes with twelve brushes — choose the 14-inch long brush handle kit for standard carboys, or the 34-inch long handle kit for heavy-duty carboys. Both brush types feature curved nylon bristles to help clean corners and small-diameter swivel heads to fit into carboy openings.
Twelve pairs of reusable cleaning gloves are right- or left-handed, unlined for easy on/off, and made of nitrile for excellent chemical resistance and durability. The long 13-inch length provides additional user protection.
Micro-90® cleaner is low foaming, rinses clean, and removes biological materials, including protein, from all types of plastics, metals, glassware, rubber, and fabric.