Cole-Parmer CFDR60-3 3/8" NPT(F) Filter Regulator (Compressor Filters/Regulators)
Representative image only.
Cole-Parmer Item # EW-79160-20
Ideal for MRO or lab applications such as cleaning compressed air or lubricating air tools.
  • Available as one-stage filter regulator or two-stage air regulator/lubricator.
  • Auto-drain valves keep water out of your system with minimal oversight.
  • Coalescent filters are provided with a differential gauge to indicate when filter needs to be changed.
  • Use connector kit to connect regulator, lubricator, or filter to your system instead of pipe nipples or pipe thread.
  • 7 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Port Size3/8\" NPT(F)
  • Thread Size3/8\" NPT(F)
  • Description3/8\" NPT(F) Filter Regulator (Compressor Filters/Regulators)
  • Warranty7 Years
Key Features
  • Available as one-stage filter regulator or two-stage air regulator/lubricator.
  • Auto-drain valves keep water out of your system with minimal oversight.
  • Coalescent filters are provided with a differential gauge to indicate when filter needs to be changed.
  • Use connector kit to connect regulator, lubricator, or filter to your system instead of pipe nipples or pipe thread.
  • 7 year warranty