Cole-Parmer Chromatography Research Supply Flame Detector Cleaning Kit
Representative image only.
Cole-Parmer Item # EW-01996-00
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionChromatography Research Supply Flame Detector Cleaning Kit
More About this Item
Kit includes a set of jet reamers, two wire minibrushes, and a dual-ended handle for reamers and brushes.
Shpg wt 1 lb (0.5 kg)