Cole-Parmer Class 1 Ductless Workstation, 18" Deep, 230V

Representative image only.
Cole-Parmer Item # EW-33730-24
Portable, compact workstations are completely self-contained
  • Integral recessed work surface contains spills
  • Top airflow is 100 CFM and has high, medium and low settings
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)12
  • Exterior Width (in)12
  • Exterior Height (in)14.5
  • Exterior Depth (in)18
  • Interior Width (in)11
  • Power (VAC)230
  • Filter TypeCarbon and particulate pre-filter
  • DescriptionClass 1 Ductless Workstation, 18\" Deep, 230V
  • Warranty1 Year
1 Year WarrantyUL Listed.

Key Features

  • Integral recessed work surface contains spills
  • Top airflow is 100 CFM and has high, medium and low settings
  • 1 year warranty

More About this Item

Compact benchtop workstations are ideal for your general chemistry, tissue research, forensic applications, and academic demonstrations. The prefilter and activated carbon filter remove fumes, odors, and nonhazardous chemicals.

Clear acrylic enclosure lets you easily view workstation contents from three sides. Front cutaways allow ergonomic use.

Larger workstations 33730-26 and -28 have a hinged sash that enables you to have a completely open front.

Filters for both size workstations are easily replaced by removing the prefilter and then unclipping the carbon filter from the rear baffle. Workstations 33730-26 and -28 feature a filter change light, but it is recommended filters be changed every 60 to 90 days with normal use. Models 33730-22 and -24 have an electrical port exit on the left side. Models 33730-26 and -28 have electrical port exits on both sides of workstation.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat