Cole-Parmer COElectrode "H" length: 300 mm
Representative image only.
Cole-Parmer Item # EW-29000-15
Measure carbon dioxide
  • In-situ-sterilizable
  • Ideal even for small-volume fermentors
  • Special internal pH electrode lets you calibrate without using gases
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes
  • Length (cm)30
  • DescriptionCOElectrode \"H\" length: 300 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • In-situ-sterilizable
  • Ideal even for small-volume fermentors
  • Special internal pH electrode lets you calibrate without using gases
  • 1 year warranty
More About this Item
For use with Cole-Parmer® ion-selective meters.