Cole-Parmer Comb for High-Throughput Horizontal Gel System; 27 Wells, 1.0 mm Thick
Representative image only.
Cole-Parmer Item # EW-28562-51
Optimize performance of your High-Throughput Horizontal Electrophoresis Gel System
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)1
  • Number of Wells27
  • DescriptionComb for High-Throughput Horizontal Gel System; 27 Wells, 1.0 mm Thick
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
These combs are compatible with your Cole-Parmer 28562-00 system. Combs are single-sided and height-adjustable; available in three thicknesses to accommodate different sample volumes. The 27- and 28-well combs are spaced to accept samples loaded using multi-channel pipettors. The 27-well combs include three molecular weight marker lanes with two spacings of 12 sample lanes; 28-well combs include four molecular weight marker lanes with three spacings of eight sample lanes.