Cole-Parmer Comb for Horizontal Mid-Size Gel System; 27 Wells, 1.5 mm Thick
Representative image only.
Cole-Parmer Item # EW-28560-59
$66.50USD / Each
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$154.00 - $3,156.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)1.5
  • Number of Wells27
  • DescriptionComb for Horizontal Mid-Size Gel System; 27 Wells, 1.5 mm Thick
More About this Item
Combs are single sided and height adjustable; available in three thicknesses to accommodate different sample volumes. The 13- and 27- well combs are spaced to allow sample loading with multi-channel pipettors.