Cole-Parmer Compact low-flow switch for gases; SPST normally open, 25 cfh/60 cfh
Representative image only.
Cole-Parmer Item # EW-32778-30
$160.70USD / Each
$296.70USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)212
  • Max Operating Temperature (° C)100
  • DescriptionCompact low-flow switch for gases; SPST normally open, 25 cfh/60 cfh
More About this Item
These normally open switches are specifically designed for reliable operation in clean air and water applications. Select from gas or liquid flow switches. Use a 50-µm filter to protect your switch. Switches include 18" (45.7 cm) L leads.