Cole-Parmer Compound Pressure Transmitter -14.7 to 100 psi; 4-20 mA Output
Representative image only.
Cole-Parmer Item # EW-68075-08
Be more accurate, affordably
  • Suitable for high-shock/vibration applications
  • SS sensor attached to SS electrode forms a variable capacitor
PRODUCT OPTIONS
Call For Price
$175.00USD / Each
for an additional
$175.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy0.25% FS
  • Min Pressure (PSI)-14.7
  • Max Pressure (PSI)100
  • Output4-20 mA
  • DescriptionCompound Pressure Transmitter -14.7 to 100 psi; 4-20 mA Output
Key Features
  • Suitable for high-shock/vibration applications
  • SS sensor attached to SS electrode forms a variable capacitor
More About this Item
Economical, excellent accuracy, and high performance. Ideal for industrial OEM equipment, engines, hydraulic systems, and compressor control.
Transducers with 4 to 20 mA output have red (positive power) and black (output) wires. Transducers with 0.5 to 5.5 V output have red (positive power), green (output), and black/white (common) wires.