Cole-Parmer Conductivity Cell, twist-lock, K=1.0
Representative image only.
Cole-Parmer Item # EW-19500-29
Ideal for by-pass streams, sample systems, and flow through applications
  • Ten-foot cables with 4-pin Military detachable connector
  • Built-in 100 Pt RTD
PRODUCT OPTIONS
$350.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 1.0
  • Cell TypeContact
  • Electrode Material316 SS, PEEK
  • Body MaterialRyton® PPS
  • Fitting1" NPT(M)
  • Mounting TypeThreaded and Twist-Lock
  • Min temperature (° C)-5
  • Max temperature (° C)135
  • Min temperature (° F)23
  • Max temperature (° F)275
  • Length (cm)15.2
  • Length (in)6
  • Cable Length (m)3
  • Cable Length (ft)10
  • DescriptionConductivity Cell, twist-lock, K=1.0
Key Features
  • Ten-foot cables with 4-pin Military detachable connector
  • Built-in 100 Pt RTD
More About this Item
Rugged twist-lock conductivity cells feature a 12-mm sensor and automatic temperature compensation element permanently sealed in a Ryton®PPS housing. Two external Viton® O-rings protect against leakage, and locking pins ensure secure positioning. Ribbed-barrel design provides easy handling. The 1" NPT(M) top thread allows for attachment of conduit.
Use the Twist-Lock sensor with adapter (listed below) for conversion of a standard pipe tee to a flow through sensor aqssembly. The optional cable provides an additional 20 feet (6 meters) of coverage.