This item is now discontinued. We recommend the following as suggested replacements: EW-78903-90
Cole-Parmer Cordless Compound Microscope, Achromatic objectives
Representative image only.
Cole-Parmer Item # EW-48922-70
Rechargeable NiMH batteries provide up to 50 hours of operation on an 8-hour charge
  • LED illumination with intensity control draws little power and provides heat-free lighting
  • Achromatic objectives provide a better alignment of the image colors for an improved image
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification40x/100x/400x/1000x
  • Maximum Magnification1000x
  • Head ConfigurationBinocular
  • ObjectivesAchromatic
  • Stage Dimensions121 x 121 mm
  • Illuminator(s)LED
  • PowerRechargeable batteries and VAC adapter
  • Power (VAC)110 to 240
  • Power (Hz)50/60
  • DescriptionCordless Compound Microscope, Achromatic objectives
  • WarrantyLTD Life Time
Canadian Standards AssociationLimited Lifetime Warranty
Key Features
  • LED illumination with intensity control draws little power and provides heat-free lighting
  • Achromatic objectives provide a better alignment of the image colors for an improved image
  • Limited Lifetime Warranty
More About this Item
Ease of use and precision optics make these microscopes ideal for the student and the professional. Binocular Siedentopf viewing head is inclined at 30° and rotates 360° for easy sharing. Objective turret is ball-bearing mounted for smooth magnification changes; reverse mounting allows easy access to specimens and stage.
Rechargeable NiMH batteries allow you to use this microscope in the field or where conventional power isn't convenient. Included recharger shuts off when batteries are fully charged, eliminating overcharging that can shorten battery life. Microscopes can be used during recharging, or recharger can be used as a power cord for 115 VAC operation.
Microscope features coarse and fine adjustments, widefield 10x eyepieces with pointer, 4-3/4" x 4-3/4" (121 x 121 mm) stage with specimen holder, and Abbe condenser with iris diaphragm.