Cole-Parmer Cordless Stereo Microscope, 20x/40x magnification
Representative image only.
Cole-Parmer Item # EW-03904-15
Compact size and cordless operation make these microscopes ideal for field use
  • Versatile top and bottom lights illuminate your sample
PRODUCT OPTIONS
$344.10USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Magnification20x/40x
  • Maximum Magnification40x
  • Head ConfigurationBinocular
  • Working Distance75 mm
  • Illuminator(s)LED, top and bottom
  • PowerRechargeable batteries with AC adapter
  • DescriptionCordless Stereo Microscope, 20x/40x magnification
Includes
transparent plate, white plate, black plate, dust cover, and rechargeable batteries with AC adapter/charger.
Key Features
  • Versatile top and bottom lights illuminate your sample
More About this Item
Compact microscopes adjust from 12" to 13-5/8" (30.5 to 34.6 cm) tall — they fit in tight spots and are easy to move where you need them. Cordless design allows field use and use in lab situations where an outlet isn't convenient. Dual-power microscopes feature parfocalled, parcentered, and achromatic objective lenses. Illumination control permits use of top and bottom lights together, top only, or bottom only.
Working distance is 75 mm and maximum specimen height is 35 mm. Stage is supplied with one transparent plate, one white plate, and one black plate for contrast viewing. Microscopes feature fixed arm, fixed head, and rugged cast-metal frame.