Cole-Parmer CPVC Submersible pH Electrode Holder, 100 Ohms RTD
Representative image only.
Cole-Parmer Item # EW-05725-42
Feature built-in ATC element and solution ground
  • A variety of housing styles available to meet your application requirements
  • Maximum pressure is 100 psi
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode Body MaterialCPVC
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorTinned ends
  • ATC Element100 Ω Pt RTD
  • DescriptionCPVC Submersible pH Electrode Holder, 100 Ohms RTD
  • Warranty1 year
1 Year Warranty
Key Features
  • A variety of housing styles available to meet your application requirements
  • Maximum pressure is 100 psi
  • 1 year warranty
More About this Item
These unique holders feature an integrated 316 stainless steel pin ground wire that serves as a temperature compensator and a solution ground. Choose economical CPVC housing or chemical-resistant KYNAR housing. In-line holders have a 1'' NPT(M) connection. Submersible holders have a 3/4'' NPT(F) thread--connect plastic pipe to match your required submersion depth.
Note: Models 05725-40 and -50 do not come with ATC elements and do not solution ground.