Cole-Parmer Cylindrical Tank with Cover, HDPE, Black, 200 Gal
Representative image only.
Cole-Parmer Item # EW-06970-10
Sturdy design complies with with FDA 21 CFR 117.1520
  • Contain drips with stepped-flange design
  • Get easier sampling of liquids—tanks over 30 gallons are designed for near-total drainage
  • No seperate part number for lid—all tanks include lid of same thickness
  • Rated for use with 1.8 specific gravity media
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • Wall thickness (in)0.25
  • Volume (Gal)200
  • Volume (Liters)757
  • Height (in)51
  • Height (cm)129.54
  • Outside Diameter (OD) (in)36
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • DescriptionCylindrical Tank with Cover, HDPE, Black, 200 Gal
  • Warranty1 Year
1 Year Warranty
Key Features
  • Contain drips with stepped-flange design
  • Get easier sampling of liquids—tanks over 30 gallons are designed for near-total drainage
  • No seperate part number for lid—all tanks include lid of same thickness
  • Rated for use with 1.8 specific gravity media
  • 1 year warranty
More About this Item
High-density polyethylene (HDPE) tanks come with or without a needle-type polypropylene spigot. Spigot has 1-1/8"-12 female screw thread. Spigot is mounted on a welded polyethylene boss and accepts 5/8" ID tubing.
Polypropylene (PP) tanks offer good stress-crack and chemical resistance. Maximum service temperature up to 104°C (220°F).
Black HDPE Tanks are manufactured with a pigmentation that protects the tank from UV light and can be used outdoors without deterioration from sunlight. Black color protects photo-sensitive media.