Cole-Parmer Dehydrated Culture Media, Azide Dextrose broth, 500 g
Representative image only.
Cole-Parmer Item # EW-14202-00
Media cultures are tested and standardized to ensure consistent and reliable performance
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
$93.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (g)500
  • DIFCO NameAzide Dextrose (0387)
  • BBL NameAzide Dextrose (11000)
  • DescriptionDehydrated Culture Media, Azide Dextrose broth, 500 g
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Use for detection of fecal Streptococci in water and sewage.
Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service.
Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.