Cole-Parmer Detachable cable; BNC, 10 ft (Electrode Cable)
Representative image only.
Cole-Parmer Item # EW-05661-09
Easily detachable electrode cable
  • Optimize performance electrodes with optional cables
  • 1 year warranty
PRODUCT OPTIONS
$85.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionDetachable cable; BNC, 10 ft (Electrode Cable)
  • Warranty1 year
1 Year Warranty
Key Features
  • Optimize performance electrodes with optional cables
  • 1 year warranty
More About this Item
Coose from BNC or stripped wire cables. Stripped end cables are used to connect the electrodes to meters, controllers and transmeters that accept bare wire connections. Stripped end cables are not for temperature compensated use.
BNC cables are used to connect the electrodes to meters, controllers and transmeters that accept BNC connections (most common).