Cole-Parmer Diamond Grip Nitrile Gloves, Size 8, Single-Layer; 12 Pair/Bg
Representative image only.
Cole-Parmer Item # EW-40216-02
Offers superior wet and dry grip
  • Hypoallergenic
  • USDA compliant
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)11
  • MaterialNitrile
  • Size8
  • Length (in)12
  • Length (cm)30.5
  • ColorGreen
  • ComplianceFDA 21 CFR 177.2600; ISO 9002
  • SterileNo
  • DescriptionDiamond Grip Nitrile Gloves, Size 8, Single-Layer; 12 Pair/Bg
Conformite Europeenne
Key Features
  • Hypoallergenic
  • USDA compliant
More About this Item
Embossed diamond pattern provides superior wet and dry grip. Latex-free nitrile gloves resist punctures, cuts, snags, and abrasion. Silicone free—suitable for use in paint and powder coat applications. Hypoallergenic. Compliance: USDA accepted per 21CFR 177.2600 for food and drug handling, manufactured under ISO 9002 standards.