Cole-Parmer Dichloromethane, ACS Grade, 4 L

Representative image only.
Cole-Parmer Item # EW-88403-53
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$250.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDichloromethane, DCM, Methylene Chloride
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingAmber Glass Bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Hazard SymbolFlammable, Toxic
  • CAS Number75-09-2
  • UN NumberUN1593
  • Shelf Life (days)60
  • DescriptionDichloromethane, ACS Grade, 4 L

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat