Cole-Parmer Essentials Lab Inoculating Needles, Sterile, Polystyrene, 1 µL, Bags of 25; 1000/Cs
Representative image only.
Cole-Parmer Essentials Item # EW-06231-09
Sterile lab inoculating needles save time and eliminate cross-contamination when transferring microbial organisms
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface for easier attachment of cells
PRODUCT OPTIONS
$82.00USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Overall Length (mm)228
  • Overall Length (in)8 31/32
  • Contained Volume (µL)1
  • ColorWhite
  • MaterialPolystyrene
  • DescriptionLab Inoculating Needles, Sterile, Polystyrene, 1 µL, Bags of 25; 1000/Cs
Sterile
Key Features
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface for easier attachment of cells
More About this Item
Ideal for picking single colonies, these disposable polystyrene lab inoculating needles are vacuum-gas plasma surface treated to allow culture medium to attach. The long, smooth, flexible stem can be bent allowing access to narrow or small-volume tubes and dishes.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these radiation sterilized (SAL 10-6) inoculating needles are available in individual peel-to-open packs, or in resealable packs of 10 or 25. Every package is printed with a lot number for quality traceability. DNase/RNase free and nonpyrogenic.