Cole-Parmer Disposable Syringe, Centric Tip, Luer Slip, 50 mL, 30/Pk
Representative image only.
Cole-Parmer Item # EW-07945-13
Ideal for all syringe filter applications
  • Constructed of durable polypropylene and polyethylene
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeCentric, Luer-Slip
  • Syringe Capacity (mL)50
  • Needle IncludedNo
  • MaterialPolypropylene and Polyethylene
  • SterileNo
  • DescriptionDisposable Syringe, Centric Tip, Luer Slip, 50 mL, 30/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Constructed of durable polypropylene and polyethylene
  • 3 month warranty
More About this Item
These all-plastic disposable syringes easily dispense small volumes of liquids. Polypropylene barrel and polyethylene plunger eliminate the need for rubber or synthetic gaskets. No silicone or oil lubricant is required in the syringe barrel. Choose either luer-slip or luer-lock connection type.