Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 25-ft Cable
Representative image only.
Cole-Parmer Item # EW-27013-45
Use for many applications in a variety of industrial housings
  • Purchase a separate housing and simply replace electrodes as needed
  • Dome bulb offers better protection from breakage
  • Stable in low-ionic strength solutions
  • Offers resistance to sodium ion error
  • Compatible with most systems
  • 1 year warranty
PRODUCT OPTIONS
$289.00USD / Each
BUY MORE & SAVE
  • 1-5$289.00USD / Each
  • 6-11$260.10USD / Each
  • 12-23$245.65USD / Each
  • 24+$231.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsWater/wastewater, plating baths
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • Max Pressure (PSI)100
  • ConnectorBNC
  • Diameter (mm)12
  • Cable Length (m)7.6
  • DescriptionDouble-Junction ORP Electrode, Gold Sensor, 25-ft Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Purchase a separate housing and simply replace electrodes as needed
  • Dome bulb offers better protection from breakage
  • Stable in low-ionic strength solutions
  • Offers resistance to sodium ion error
  • Compatible with most systems
  • 1 year warranty
More About this Item
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).