Cole-Parmer Draft Shield, for 128 x 128 mm Weighing Pan
Representative image only.
Cole-Parmer Item # EW-10100-93
  • For Cole-Parmer Touchscreen Series Balances with weighing pan size of 128 x 128 mm
  • 1 month warranty
PRODUCT OPTIONS
$312.50USD / Each
$334.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeDraft Shield
  • DescriptionDraft Shield, for 128 x 128 mm Weighing Pan
1 Month Warranty
Key Features
  • For Cole-Parmer Touchscreen Series Balances with weighing pan size of 128 x 128 mm
  • 1 month warranty
More About this Item
Replacement parts and accessories for your Cole-Parmer Basic and Advanced Touch-Screen Balances.
Obtain more stable readings with small samples—protects sample from air drafts