Cole-Parmer Dual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm

Representative image only.
Cole-Parmer Item # EW-28575-00
Customize your own gel system depending on your needs and budget!
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)250
  • Cell Width (in)6.67
  • Cell Height (in)5
  • Cell Depth (in)7.75
  • Cell Width (cm)16.94
  • Cell Height (cm)12.7
  • Cell Depth (cm)19.69
  • Glass Width (in)4.45
  • Glass Width (cm)11.3
  • Glass Height (cm)10
  • DescriptionDual Vertical Mini-Gel System, gel bed dimensions 10 x 10 cm
  • Warranty3 year
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Mini Dual Gel Units. Small format is ideal for mini SDS-PAGE and mini agarose gel separations, plus it reduces the use of expensive reagents. These dual units let you run two agarose or polyacrylamide gels simultaneously under identical conditions; also compatible with most precast gels. The built-in cooling system ensures fast run times and flat, even banding patterns. Chamber gasket and clamps create a positive, leakproof seal.

Required System Components:
1) Gel Unit (order power supply separately)

For the Gel Wrap™ Gasket gel casting method, you will need:
2) Glass plate set(s)
3) Casting clamps (three per plate set for mini units)

Order the same thickness of:
4) GelWrap gasket (one per plate set)
5) Spacer set (one per plate set)
6) Combs (one per plate set)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat