Cole-Parmer Economical Orbital Shaker; 12" x 12" Platform, 110-240 VAC (Reconditioned)
Representative image only.
Cole-Parmer Item # EW-00168-VN
Low cost – incorporates features typically found in more expensive models
  • Sealed motor ensures quiet, trouble-free operation
  • 50-250 rpm
Call For Price
SPECS & DESCRIPTION
Specifications & Description
Includes
6-ft cord with U.S. standard plug.
Key Features
  • Sealed motor ensures quiet, trouble-free operation
  • 50-250 rpm
More About this Item
Microprocessor based keypad with digital display tracks preset timed run, continuous run, and rpm settings — improves repeatability of test procedures. Skid-free black neoprene sheeting provides a nonslip surface for petri dishes, culture bottles, and flasks. Continually adjustable speed lets you obtain gentle agitation or vigorous orbital motion. Slow start and stop motion decreases risk of spills. Anodized aluminum platform provides a sturdy surface for samples.
Call Us At1-847-549-7600
Live chat