Cole-Parmer EDTA, Disodium Dihydrate, ACS Grade, 1000 g

Representative image only.
Cole-Parmer Item # EW-88403-57
Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs

$188.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaC10H14N2O8Na2•2H2O
  • ClassReagent Salts
  • ChemistryAcids
  • FormSolid
  • SynonymsEthylenediaminetetraacetic Acid
  • Weight (g)1000
  • Formula Weight372.24
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number6381-92-6
  • Shelf Life (days)60
  • DescriptionEDTA, Disodium Dihydrate, ACS Grade, 1000 g

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat