Cole-Parmer Electronic Multichannel Pipette, 12 Channel, 5 to 100 ul
Representative image only.
Cole-Parmer Item # EW-24500-29
Multiple modes meet your pipetting needs
  • Preprogrammed with six different operations
  • Adjustable aspiration and pipetting speeds
  • Large color LED display is easy to read
  • Lightweight, ergonomic, and simple to use
  • Available with 12 channels
PRODUCT OPTIONS
$1,270.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes - tip cone
  • Max Volume (µL)100
  • Min Volume (µL)5
  • Increments (µL)0.1
  • Accuracy (%)±2.0
  • Reproducibility±1.0%
  • Number Of Channels12
  • Tip EjectorYes
  • Auto ShutoffYes
  • CalibrationInternal
  • BatteryLithium ion
  • DescriptionElectronic Multichannel Pipette, 12 Channel, 5 to 100 ul
Includes
Li-ion battery and charger.
Key Features
  • Preprogrammed with six different operations
  • Adjustable aspiration and pipetting speeds
  • Large color LED display is easy to read
  • Lightweight, ergonomic, and simple to use
  • Available with 12 channels
  • Volume range of 5 to 100 µL
More About this Item
The electronic multichannel pipettors feature a large easy-to-read color LED glass display and a simple to use menu. Multiple modes of pipetting are available: forward, gel loading, manual, mixing, reverse, sample mixing, sub-sample, titration, and unequal volume.
You can program the pipettor with your own protocol or choose from one of the six preprogrammed operations. The pipettors can be used with a variety of liquids including easy-foaming, high-viscosity, and volatile liquids. Other features include soft tip ejection, internal automatic calibration, adjustable aspiration, and autoclavable tip cone. The entire pipettor can also be sterilized by UV light.
Choose from 8- or 12-channel pipettors to meet your laboratory needs.