Cole-Parmer Electrophoretic Blotting System, 3 Cassette capacity

Representative image only.
Cole-Parmer Item # EW-28580-00
For Western, Southern, or Northern blots – transfer proteins and nucleic acids from agarose or acrylamide gels
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)3800
  • Cell Width (in)4.3
  • Cell Height (in)11.5
  • Cell Depth (in)7.5
  • Cell Width (cm)10.92
  • Cell Height (cm)29.21
  • Cell Depth (cm)19.05
  • DescriptionElectrophoretic Blotting System, 3 Cassette capacity
  • Warranty3 Years
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Glass cooling chamber with tubing adapters is built into base for temperature-controlled runs. Platinum wire electrodes are mounted in recessed grooves for protection from damage when cleaning unit or loading cassettes. Electrode design provides even current distribution over entire gel surface. Place a magnetic stirring bar beneath cassettes to circulate buffer within unit for more efficient cooling. Order additional cassettes separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat