Cole-Parmer Epoxy body, single junction ORP electrode
Representative image only.
Cole-Parmer Item # EW-59001-86
Durable and low-maintenance electrodes
  • Ideal for laboratory and field applications
  • Epoxy-body housing resists impact
  • Gel-filled, seladed body is low maintenance
  • Compatible with most brands of meters
PRODUCT OPTIONS
$69.30USD / Each
$77.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeORP
  • Electrode Body MaterialEpoxy
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionEpoxy body, single junction ORP electrode
Key Features
  • Ideal for laboratory and field applications
  • Epoxy-body housing resists impact
  • Gel-filled, seladed body is low maintenance
  • Compatible with most brands of meters
More About this Item
Epoxy-body electrodes are a selaled, gel-filled design requiring no maintenance. Single-junction design has a Ag/AgCl reference. Dimensions are 12.5 mm diameter x 90 mm length.