Cole-Parmer Essentials Flat cover for 07237-10, 07239-10, 07240-10, 07241-00
Representative image only.
Cole-Parmer Essentials Item # EW-07240-60
Reduce evaporation
  • 10-32 coned port
  • 1.9 µL swept volume
  • Assembly includes SS nut and washer
  • 1 year warranty
PRODUCT OPTIONS
$97.00USD / Each
BUY MORE & SAVE
  • 1-5$97.00USD / Each
  • 6-29$87.30USD / Each
  • 30-119$82.45USD / Each
  • 120+$77.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlat cover for 07237-10, 07239-10, 07240-10, 07241-00
  • Warranty1 year
1 Year Warranty
Key Features
  • 10-32 coned port
  • 1.9 µL swept volume
  • Assembly includes SS nut and washer
  • 1 year warranty
More About this Item
These stainless steeel covers help reduce evaporation, while containing harmful vapors and keeping contaminants out.