Cole-Parmer Essentials Graduated Cylinder, PP 10 mL; 2/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06134-61
Don’t guess at volume
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
$15.50USD / Pkg of 2
BUY MORE & SAVE
  • 1-5$15.50USD / Pkg of 2
  • 6-29$13.95USD / Pkg of 2
  • 30-119$13.18USD / Pkg of 2
  • 120+$12.40USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Capacity (mL)10
  • Graduation Interval (mL)0.2
  • DescriptionGraduated Cylinder, PP 10 mL, 2/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
More About this Item
These polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability. Cylinders have raised graduations for easy reading. Autoclavable.