Cole-Parmer Essentials Graduated Cylinder, PP 25 mL; 12/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06134-64
Don’t guess at volume
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Capacity (mL)25
  • Graduation Interval (mL)0.5
  • DescriptionGraduated Cylinder, PP 25 mL, 12/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
More About this Item
Bulk packed for greater savings, these polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability, these cylinders have raised graduations for easy reading. Autoclavable.