Cole-Parmer Essentials Graduated Cylinder, PP 250 mL; 6/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06134-72
Don’t guess at volume
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
PRODUCT OPTIONS
$67.00USD / Pkg of 6
BUY MORE & SAVE
  • 1-5$67.00USD / Pkg of 6
  • 6-29$60.30USD / Pkg of 6
  • 30-119$56.95USD / Pkg of 6
  • 120+$53.60USD / Pkg of 6
$81.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Capacity (mL)250
  • Graduation Interval (mL)2
  • DescriptionGraduated Cylinder, PP 250 mL, 6/pk
Key Features
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
More About this Item
Bulk packed for greater savings, these polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability, these cylinders have raised graduations for easy reading. Autoclavable.